Learn More
GFP Rat, Unconjugated, Clone: 3H9, Proteintech ChromoTek
Description
Rat monoclonal [3H9] to GFP
Specifications
Specifications
| Antigen | GFP |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 3H9 |
| Conjugate | Unconjugated |
| Host Species | Rat |
| Quantity | 20 μL |
| Primary or Secondary | Primary |
| Content And Storage | Shipped at ambient temperature. Store at +4°C. Stable for one year. |
| Isotype | IgG2a |