Learn More
GFP Rat, Unconjugated, Clone: 3H9, Proteintech ChromoTek
Rat Monoclonal Antibody
Brand:  Chromotek 3H9-100
 View more versions of this product
Product Code. 17363263
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Description
Rat monoclonal [3H9] to GFP
Specifications
Specifications
GFP
Monoclonal
Unconjugated
100 μL
Shipped at ambient temperature. Store at +4°C. Stable for one year.
Western Blot
3H9
Rat
Primary
IgG2a