missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFP Rabbit, Unconjugated, Polyclonal, Proteintech ChromoTek
Description
Rabbit polyclonal antibody to Green Fluorescent Protein
Specifications
Specifications
| Antigen | GFP |
| Applications | Western Blot, Immunofluorescence, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Quantity | 20 μL |
| Primary or Secondary | Primary |
| Content And Storage | Shipped on blue ice. Store at +4°C short term (1-2 weeks). Aliquot upon delivery. Store at -20°C or -80°C long term. Avoid freeze-thaw cycles. Stable for one year. |
| Isotype | IgG2a |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?