missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFP Rabbit anti-Nonspecies specific, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
204.00€
Specifications
| Antigen | GFP |
|---|---|
| Dilution | Western Blot 1:1000 - 1:5000, Immunocytochemistry/Immunofluorescence 1:2000 - 1:5000 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
GFP Polyclonal specifically detects GFP in Non-species specific samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| GFP | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Epitope Tags | |
| 50% PBS and 50% Glycerol | |
| This GFP antibody was developed against full length recombinant GFP. | |
| Primary |
| Western Blot 1:1000 - 1:5000, Immunocytochemistry/Immunofluorescence 1:2000 - 1:5000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Non-species specific | |
| Enhanced Green Fluorescent Protein, Green Fluorescent Protein, green fluorescent protein (gfp) | |
| IgG | |
| Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title