Learn More
Abnova™ GFER (Human) Recombinant Protein
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Description
- Sequence: MRTQQKRDTKFREDCPPDREELGRHSWAVLHTLAAYYPDLPTPEQQQDMAQFIHLFSKFYPCEECAEDLRKRLCRNHPDTRTRACFTQWLCHLHNEVNRKLGKPDFDCSKVDERWRDGWKDGSCD
Specifications
Specifications
| Accession Number | AAH28348 |
| Gene ID (Entrez) | 2671 |
| Name | growth factor, augmenter of liver regeneration |
| Preparation Method | Wheat germ expression system |
| Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
| Quantity | 25 μg |
| Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Gene Alias | ALR, ERV1, HERV1, HPO, HPO1, HPO2, HSS |
| Gene Symbol | GFER |
| Species | Wheat Germ (in vitro) |
| Show More |