missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFAP Antibody (CL2713) [DyLight 550], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-76477R
This item is not returnable.
View return policy
Description
GFAP Monoclonal antibody specifically detects GFAP in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown
Specifications
| GFAP | |
| Monoclonal | |
| DyLight 550 | |
| ALXDRD, FLJ45472, GFAP, GFAP astrocytes, glial fibrillary acidic protein | |
| This antibody was developed using a recombinant protein derived from P14136, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Astrocyte Markers, Cancer, Cell Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Inflammation, Neurodegeneration, Neuroscience, Signal Transduction, Stem Cell Markers | |
| 2670 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown | |
| CL2713 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction