Learn More
Thermo Scientific Chemicals Gentiopicroside
Product Code. 17291272 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17291272 250 mg 250mg
17281272 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17291272 Supplier Thermo Scientific Chemicals Supplier No. 466832500
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 20831-76-9
Molecular Formula C16H20O9
Specifications
Quantity 250 mg
Melting Point 191.0°C
Formula Weight 356.33
InChI Key DUAGQYUORDTXOR-GPQRQXLASA-N
SMILES OC[C@H]1O[C@@H](O[C@@H]2OC=C3C(=O)OCC=C3[C@H]2C=C)[C@H](O)[C@@H](O)[C@@H]1O
IUPAC Name (5R,6S)-5-ethenyl-6-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-1H,3H,5H,6H-pyrano[3,4-c]pyran-1-one
Molecular Weight (g/mol) 356.33
Chemical Name or Material Gentiopicroside
compliance-icons
Product Identifier
  • Gentiopicroside
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Gentiopicroside