accessibility menu, dialog, popup

UserName

CAS RN 20831-76-9

OH(R)O(S)O(S)OOO(R)H2C(R)OH(S)OH(S)HO

CAS RN 20831-76-9

IUPAC Name: (5R,6S)-5-ethenyl-6-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-1H,3H,5H,6H-pyrano[3,4-c]pyran-1-one
Synonyms: gentiopicroside gentiopicrin
Molecular Weight (g/mol): 356.33
Molecular Formula: C16H20O9
InChi Key: DUAGQYUORDTXOR-GPQRQXLASA-N
SMILES: OC[C@H]1O[C@@H](O[C@@H]2OC=C3C(=O)OCC=C3[C@H]2C=C)[C@H](O)[C@@H](O)[C@@H]1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results