Learn More
General Purpose Viscosity Standard, N14, Paragon Scientific™
Product Code. 15351214
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15351214 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15351214 Supplier Paragon Scientific Supplier No. N14
  • ONLINE EXCLUSIVE PRICE
    103.05€
    100.80€ / 500mL
    Save 2.25€
    2% Off
 Restricted Item
In Stock
Add to basket
Edge
Specifications
Viscosity 30.3mm2/sec. (cSt) at 20°C Kinematic Viscosity
Quantity 500 mL
compliance-icons compliance-icons
Product Identifier
  • N14
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Aspiration toxicity Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P331-Do NOT induce vomiting.