missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GDNF Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
369.00€ - 559.00€
Specifications
| Antigen | GDNF |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18294254
|
Novus Biologicals
NBP2-56036 |
100 μL |
559.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18615687
|
Novus Biologicals
NBP2-56036-25ul |
25 μL |
369.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GDNF Polyclonal specifically detects GDNF in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| GDNF | |
| Polyclonal | |
| Rabbit | |
| Apoptosis, Cancer | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 2668 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:EDRSLGRRRAPFALSSDSNMPEDYPDQFDDVMDFIQATIKRLKRSPDKQMAVLPRRERNRQAAAANPENSRG | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| Astrocyte-derived trophic factor, ATF, ATF1, ATF2, glial cell derived neurotrophic factor, glial cell line derived neurotrophic factor, glial cell line-derived neurotrophic factor, HFB1-GDNF, hGDNF, HSCR3 | |
| GDNF | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title