missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axin-1 Antibody (4E9F1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61695-0.025ml
This item is not returnable.
View return policy
Description
Axin-1 Monoclonal specifically detects Axin-1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Axin-1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:50-1:250 | |
| axin 1, axin-1, AXINaxis inhibitor 1, Axis inhibition protein 1, fused, mouse, homolog of, hAxin, MGC52315 | |
| Mouse | |
| 95.6 kDa | |
| 0.025 mL | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG2b |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| 4E9F1 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| AXIN1 | |
| Purified recombinant fragment of human AXIN1 (AA: 546-752) expressed in E. Coli. | |
| Protein G purified | |
| Apoptosis, Wnt Signaling Pathway | |
| 8312 | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction