missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GDF1 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5149483
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
Growth differentiation factors (GDFs) are members of the transforming growth factor (TGF) superfamily that is involved in embryonic development and adult tissue homeostasis. GDF1 was initially identified as a temporally expressed gene in the mouse central nervous system during embryonic development, with only one isoform detected in adult tissues. GDF1 is required for left-right patterning during development and directly interacts with Nodal, another member of the TGF-beta superfamily. It has been suggested that GDF1 regulates the activity and signaling range of Nodal through direct interaction.
Specifications
| GDF1 | |
| Polyclonal | |
| Unconjugated | |
| GDF1 | |
| AI385651; DORV; DTGA3; Embryonic growth/differentiation factor 1; Gdf1; GDF-1; growth differentiation factor 1; RAI | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 14559 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| P20863 | |
| GDF1 | |
| E. coli-derived recombinant mouse GDF-1, His238-Arg357. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction