missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
GCM1 Polyclonal antibody specifically detects GCM1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | GCM1 |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL |
| Formulation | PBS (pH 7.2), 40% Glycerol |
| Gene Alias | GCM motif protein 1, GCMA, glial cells missing (Drosophila) homolog a, Glial cells missing homolog 1, glial cells missing homolog 1 (Drosophila), hGCMachorion-specific transcription factor GCMa |
| Host Species | Rabbit |
| Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: LGRNHLADNCYSNYPFPLTSWPCSFSPSQNSSEPFYQQLPLEPPAAKTGCPPLWPNPAGNLYEEKVHVDFNSYVQSPAYHSPQEDPFLFTYASHPHQQYSLPSKSSKWDFEEEMTYLGLDHCNNDMLLNLCP |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?