Learn More
GBP4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17746-1-AP-150UL
This item is not returnable.
View return policy
Description
Guanylate-binding proteins, such as GBP4, are induced by interferon and hydrolyze GTP to both GDP and GMP.[supplied by OMIM, Dec 2008].Specifications
| GBP4 | |
| Polyclonal | |
| Unconjugated | |
| GBP4 | |
| GBP 4, GBP4, GTP binding protein 4, guanylate binding protein 4, Mpa2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 115361 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96PP9 | |
| GBP4 | |
| GBP4 Fusion Protein Ag12006 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |