missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GB 23200.100-2016 Pyrethroide Pesticide Mixture 677 100 μg/mL in Toluene:Hexane, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | (±)-Fenpropathrin∣Bifenthrin∣Cyfluthrin∣Cypermethrin∣Deltamethrin∣Fenvalerate∣lambda-Cyhalothrin∣Permethrin∣tau-Fluvalinate |
| CAS | 39515-41-8, 82657-04-3, 68359-37-5, 52315-07-8, 52918-63-5, 51630-58-1 |
| Analyte or Component Names | (±)-Fenpropathrin∣Bifenthrin∣Cyfluthrin∣Cypermethrin∣Deltamethrin∣Fenvalerate∣lambda-Cyhalothrin∣Permethrin∣tau-Fluvalinate |
| Shipping Condition | +20°C |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Hexane/Toluene |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | (±)-Fenpropathrin 100 μg/mL∣Bifenthrin 100 μg/mL∣Cyfluthrin 100 μg/mL∣Cypermethrin 100 μg/mL∣Deltamethrin 100 μg/mL∣Fenvalerate 100 μg/mL∣lambda-Cyhalothrin 100 μg/mL∣Permethrin 100 μg/mL∣tau-Fluvalinate 100 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?