Learn More
GAT3 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97671MFV450
This item is not returnable.
View return policy
Description
GAT3 Polyclonal antibody specifically detects GAT3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| GAT3 | |
| Rabbit | |
| 6538 |
| Polyclonal | |
| Produced in rabbits immunized with E. coli-derived Human GAT3 fragment. | |
| IgG |