missing translation for 'onlineSavingsMsg'
Learn More

Gasoline 20000 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30317604
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
5mL
This item is not returnable. View return policy

Product Code. 30317604

Brand: Dr. Ehrenstorfer DREYS09010020ME

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Regular Unleaded Gasoline
CAS 8006-61-9
Analyte or Component Names Regular Unleaded Gasoline
Shipping Condition Room Temperature
Concentration 20000 μg/mL
Certifications/Compliance ISO 17034
Synonym Gasoline, Gasoline, natural, Natural gasoline, Petrol, natural, Gasoline, natural (10C, 11C, 14C), Oxygenate-free Rfa Gasoline, Oxygenate Free Gasoline, Unleaded, Premium
Solvent or Matrix Methanol
Quantity 5 x 1 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.