missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAS7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35418-100ul
This item is not returnable.
View return policy
Description
GAS7 Polyclonal antibody specifically detects GAS7 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| GAS7 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| GAS-7, growth arrest-specific 7, KIAA0394growth arrest-specific protein 7, MGC1348, MLL/GAS7, MLL/GAS7 fusion protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-412 of human GAS7 (NP_001124303.1).,, Sequence:, MVPPPPGEESQTVILPPGWQSYLSPQGRRYYVNTTTNETTWERPSSSPGIPASPGSHRSSLPPTVNGYHASGTPAHPPETAHMSVRKSTGDSQNLGSSSPSKKQSKENTITINCVTFPHPDTMPEQQLLKPTEWSYCDYFWADKKDPQGNGTVAGFELLLQKQLKGKQMQKEMSEFIRERIKIEEDYAKNLAKLSQNSLASQEEGSLGEAWAQVKKSLADEAEVHLKFSAKLHSEVEKPLMNFRENFKKDMKKCDHHIADLRKQLASRYASVEKARKALTERQRDLEMKTQQLEIKLSNKTEEDIKKARRKSTQAGDDLMRCVDLYNQAQSKWFEEMVTTTLELERLEVERVEMIRQHLCQYTQLRHETDMFNQSTVEPVDQLLRKVDPAKDRELWVREHKTGNIRPVDMEI | |
| 100 μL | |
| Cell Cycle and Replication | |
| 8522 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction