Learn More
Thermo Scientific™ Gas Chromatography Tubing
Product Code. 15264603 Shop All Thermo Scientific Products
Click to view available options
Material:
Copper
Stainless Steel
Diameter (English) Inner:
0.01 in.
0.02 in.
0.03 in.
0.04 in.
0.065 in.
0.085 in.
0.19 in.
0.21 in.
Diameter (English) Outer:
1/16 in.
1/4 in.
1/8 in.
Description:
25 ft. Length
50 ft. Length
Unit Size:
Each
Product Code. 15264603
Brand: Thermo Scientific™ 60181634
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Find the copper and stainless steel tubing in a variety of sizes for plumbing your gas chromatography (GC) system.
Find copper and stainless steel Thermo Scientific™ Gas Chromatography Tubing in a variety of sizes for plumbing your gas chromatography (GC) system.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
TRUSTED_SUSTAINABILITY
Specifications
Type Gas Chromatography Tubing
Material Stainless Steel
Description 25 ft. Length
Diameter (English) Inner 0.01 in.
Diameter (English) Outer 1/16 in.
For Use With (Equipment) Gas Chromatography Systems