Learn More
Thermo Scientific™ Gas Chromatography 3-Port SilFlow Replacement Parts
Product Code. 15277488 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Ferrule
FingerTite Tool
Silflow Nuts
For Use With (Equipment):
3-Port SilFlow Device
OD 0.36 mm (ID 0.25mm) Columns
OD 0.45 mm (ID 0.32mm) Columns
Unit Size:
Each
Pack of 10
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type For Use With (Equipment) Unit Size
15277488 Silflow Nuts 3-Port SilFlow Device Pack of 10
15270339 Ferrule OD 0.36 mm (ID 0.25mm) Columns Pack of 10
15280339 Ferrule OD 0.45 mm (ID 0.32mm) Columns Pack of 10
15207518 FingerTite Tool 3-Port SilFlow Device Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15277488 Supplier Thermo Scientific™ Supplier No. 290SF302
Estimated Shipment: 03-12-2026
Switch your GC column in GC-MS without having to vent.
Maintain reliability with 3-Port SilFlow replacement parts when you place your next order.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
TRUSTED_SUSTAINABILITY
Specifications
Type Silflow Nuts
For Use With (Equipment) 3-Port SilFlow Device
Quantity 10/Pk.