missing translation for 'onlineSavingsMsg'
Learn More

GAPDH Antibody (1A10), DyLight 680, Novus Biologicals™

Product Code. 18282363 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18282363 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18282363 Supplier Novus Biologicals Supplier No. NBP147339FR
  • ONLINE EXCLUSIVE PRICE
    579.00€
    547.05€ / 0.1mL
    Save 31.95€
    5% Off
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Mouse Monoclonal Antibody

GAPDH Monoclonal specifically detects GAPDH in Human, Mouse (Negative), Rat (Negative) samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GAPDH
Applications ELISA, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1A10
Conjugate DyLight 680
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias aging-associated gene 9 protein, EC 1.2.1, EC 1.2.1.12, EC 2.6.99.-, G3PD, GAPD, glyceraldehyde 3-phosphate dehydrogenase, glyceraldehyde-3-phosphate dehydrogenase, MGC88685, Peptidyl-cysteine S-nitrosylase GAPDH
Gene Symbols GAPDH
Host Species Mouse
Immunogen GAPDH antibody developed against purified recombinant fragment of human GAPDH expressed in E. coli. [UniProt# P04406]
Purification Method Ammonium sulfate precipitation
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Apoptosis, Autophagy, Cancer, DNA Repair, Lipid and Metabolism, Loading Controls, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Nuclear Receptors Coactivators and Corepressors, Signal Transduction, Translation Control
Primary or Secondary Primary
Gene ID (Entrez) 2597
Target Species Human, Mouse (Negative), Rat (Negative)
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.