missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAP-43 Antibody (CL10847) [Allophycocyanin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-21181APC
This item is not returnable.
View return policy
Description
GAP-43 Monoclonal antibody specifically detects GAP-43 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| GAP-43 | |
| Monoclonal | |
| APC | |
| Axonal membrane protein GAP-43, B-50, calmodulin-binding protein P-57, growth associated protein 43, Growth-associated protein 43, nerve growth-related peptide GAP43, Neural phosphoprotein B-50, neuromodulin, neuron growth-associated protein 43, PP46, protein F1 | |
| This antibody was developed using a synthetic peptide derived from P17677, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer, Cellular Markers, Growth and Development, Neuronal Cell Markers, Neuroscience, Phospho Specific | |
| 2596 | |
| Store at 4⪚C in the dark. | |
| IgG2b |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL10847 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction