Learn More
Berkshire™ Gamma Wipe™ 120 Cleanroom Wipers
BIOPHARMA
Product Code. p-200022126 Shop All Berkshire Products
Click to view available options
Dimensions (L x W):
12 x 12 in.
Unit Size:
Pack of 300
Product Code. 12887143
Brand: Berkshire™ GW120ST15
Only null left
Add to basket
Add to basket
Includes
Certificate of sterility.
Gamma Wipe™ 120 is a sterile cleanroom laundered 100% polyester knit wiper.
  • Low particles, fibers, ions and extractables
  • Continuous-filament polyester knit fabric
  • High absorbency
  • High abrasion resistance
  • Soft hand and edge for sensitive surfaces
  • Chemically compatible with common cleaning and disinfecting solutions including isopropyl alcohol, phenols, bleaches and quarternary ammonium compounds
  • For use in ISO Class 4 (Class 10) applications
Order Info
Basis weight: 151 g/m2.
Compliance
Gamma irradiated and validated sterile to a Sterility Assurance Level of 10 -6 in accordance with ANSI/AAMI/ISO American National Standards and European Standards EN1174 and EN552.
Specifications
Product Type Cleanroom Wipe
Length (English) 12 in.
Length (Metric) 30.48 cm
Width (English) 12 in.
Width (Metric) 30.48 cm
Material Polyester
Absorbancy Highly Absorbent
Compatibility Common cleaning and disinfecting solutions
Autoclavable Not Autoclavable
Basis Weight 151 gsm
Certifications/Compliance RoHS 2002/05, REACH
Construction Knit
Edges Knife Cut
Dimensions (L x W) 12 x 12 in.
For Use With (Application) For use in ISO Class 4 and higher sterile environments where microbial exclusion is demanded, excellent for cleaning medical devices
For Use With (Equipment) For cleaning sterile laminar air flow units, barrier isolators and bio-containment units
Ply 1-ply