missing translation for 'onlineSavingsMsg'
Learn More
Learn More
gamma Catenin Antibody (4C12), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
217.00€ - 552.00€
Specifications
| Antigen | gamma Catenin |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18683936
|
Novus Biologicals
NBP2-37557-0.025ml |
0.025 mL |
217.00€
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18756623
|
Novus Biologicals
NBP2-37557 |
0.1 mL |
552.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
gamma Catenin Monoclonal specifically detects gamma Catenin in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| gamma Catenin | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, Cytoskeleton Markers | |
| Ascites with 0.03% Sodium Azide | |
| 3728 | |
| Purified recombinant fragment of human JUP expressed in E. Coli. | |
| Primary | |
| 82 kDa |
| Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human, Mouse | |
| catenin (cadherin-associated protein), gamma (80kD), catenin (cadherin-associated protein), gamma 80kDa, Catenin gamma, Desmoplakin III, Desmoplakin-3, DP3CTNNG, DPIII, gamma-catenin, junction plakoglobin, PDGBARVD12, PKGB | |
| JUP | |
| IgG1 | |
| Unpurified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title