missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GALR1 Monoclonal Antibody (613207)

Product Code. 30534596
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30534596 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30534596 Supplier Invitrogen™ Supplier No. MA560880

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Reconstitute at 0.5 mg/mL in sterile PBS.

The neuropeptide galanin elicits a range of biological effects by interaction with specific G-protein-coupled receptors. Galanin receptors are seven-transmembrane proteins shown to activate a variety of intracellular second-messenger pathways. GALR1 inhibits adenylyl cyclase via a G protein of the Gi/Go family. GALR1 is widely expressed in the brain and spinal cord, as well as in peripheral sites such as the small intestine and heart.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GALR1
Applications Flow Cytometry
Classification Monoclonal
Clone 613207
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with trehalose and no preservative
Gene GALR1
Gene Accession No. P47211
Gene Alias Gal receptor 1; Gal-1 receptor; GAL1-R; Galanin r1 receptor; galanin receptor 1; galanin receptor type 1; Galanin-1 receptor; Galn1r; GALNR; Galnr1; Galr1; Gal-r1; GALR-1; Hugal 1-r
Gene Symbols GALR1
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human Galanin R1/GALR1.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2587
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.