missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gallamine Triethiodide, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Gallamine Triethiodide
Specifications
Specifications
| Chemical Name or Material | Gallamine triethiodide |
| CAS | 65-29-2 |
| Analyte or Component Names | Gallamine triethiodide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C30 H60 N3 O3 . 3 I |
| Certifications/Compliance | ISO 17025 |
| Synonym | Ammonium, [v-phenenyltris(oxyethylene)]tris[triethyl-, triiodide (8CI), Ethanaminium, 2,2',2'-[1,2,3-benzenetriyltris(oxy)]tris[N,N,N-triethyl-, triiodide (9CI), [v-Phenenyltris(oxyethylene)]tris[triethylammonium iodide] (6CI,7CI), 1,2,3-Tris(2-diethylaminoethoxy)benzene triethiodide, 1,2,3-Tris(2-diethylaminoethoxy)benzene tris(ethyliodide), 1,2,3-Tris(2-triethylammonium ethoxy)benzene triiodide, 1,2,3-Tris(diethylaminoethoxy)benzene triethiodide, 3697 RP, Benzcurine iodide, F 2559, Flacedil, Flaxedil, Flaxedil iodide, Fourneau 2559, Gallaflex, Gallamin triethiodide, Gallamine iodide, Gallamine triethiodide, Gallamine triiodoethylate, HL 8583, Miowas G, Parexyl, Pyrogallol 1,2,3-(diethylaminoethyl ether) tris(ethyliodide), Pyrolaxon, RP 3697, Relaxan, Remyolan, Retensin, Sincurarine, Syncurarine, Tri(diethylaminoethoxy)-1,2,3-benzene triiodoethylate, Ethanaminium, 2,2',2'-[1,2,3-benzenetriyltris(oxy)]tris[N,N,N-triethyl-, iodide (1:3), 1,2,3-Tri(beta-diethylaminoethoxy)benzene triethiodide, Tri(beta-diethylaminoethoxy)-1,2,3-benzene tri-iodoethylate |
| InChI Formula | InChI=1S/C30H60N3O3.3HI/c1-10-31(11-2,12-3)22-25-34-28-20-19-21-29(35-26-23-32(13-4,14-5)15-6)30(28)36-27-24-33(16-7,17-8)18-9;;;/h19-21H,10-18,22-27H2,1-9H3;3*1H/q+3;;;/p-3 |
| SMILES | [I-].[I-].[I-].CC[N+](CC)(CC)CCOc1cccc(OCC[N+](CC)(CC)CC)c1OCC[N+](CC)(CC)CC |
| IUPAC Name | 2-[2,3-bis[2-(triethylazaniumyl)ethoxy]phenoxy]ethyl-triethylazanium;triiodide |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction