missing translation for 'onlineSavingsMsg'
Learn More

Galectin-9 Antibody (1015214) [Janelia Fluor™ 585], Novus Biologicals™

Product Code. 30162341 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30162341 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30162341 Supplier Novus Biologicals Supplier No. FAB20453JF585

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Galectin-9 Monoclonal antibody specifically detects Galectin-9 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen Galectin-9
Applications Flow Cytometry, Immunohistochemistry, Western Blot, CyTOF
Classification Monoclonal
Clone 1015214
Conjugate Janelia Fluor 585
Formulation 50mM Sodium Borate
Gene Alias ecalectin, Gal-9, galectin 9, galectin-9, HUAT, lectin, galactoside-binding, soluble, 9, LGALS9A, MGC117375, MGC125973, MGC125974, Tumor antigen HOM-HD-21, urate transporter/channel protein
Host Species Mouse
Immunogen Human embryonic kidney cell, HEK293 derived human Galectin-9, Met1-Thr323, Accession # NP_002299
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Myeloid derived Suppressor Cell, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3965
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.