missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galectin-3 Rabbit Recombinant Antibody, Proteintech
Rabbit Recombinant Antibody
Brand: Proteintech 82024-1-RR-20UL
This item is not returnable.
View return policy
Description
The Galectin-3 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human Galectin-3. This antibody recognizes human, mouse antigen. The Galectin-3 antibody has been validated for the following applications: WB, IHC, IF/ICC, IP, ELISA analysis.Specifications
| Galectin-3 | |
| Recombinant Monoclonal | |
| 500 μg/mL | |
| CBP35; GAL3; GALBP; GALIG; LGALS2; MAC2 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| 31 kDa | |
| Liquid |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, ELISA | |
| 4M10 | |
| Unconjugated | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 3958 | |
| Store at -20°C. Stable for one year after shipment. | |
| Recombinant Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction