missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galectin-1 Antibody (GAL1/1831), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54457AF594
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Galectin-1 Monoclonal antibody specifically detects Galectin-1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Protein Array, CyTOF
Spécification
| Galectin-1 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 14 kDa laminin-binding protein, 14 kDa lectin, Beta-galactoside-binding lectin L-14-I, DKFZp686E23103, gal-1, GAL1, Galaptin, galectin 1, galectin-1, GBP, HBL, HLBP14, HPL, Lactose-binding lectin 1, Lectin galactoside-binding soluble 1, lectin, galactoside-binding, soluble, 1, Putative MAPK-activating protein PM12, S-Lac lectin 1 | |
| Recombinant fragment (around aa12-108) of human Galectin-1 protein (exact sequence is proprietary) (Uniprot: P09382) | |
| 0.1 mL | |
| Apoptosis, BiologicAll speciesy Active Proteins | |
| 3956 | |
| Store at 4°C in the dark. | |
| IgG1 λ |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| GAL1/1831 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu