missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gag-Pro-Pol polyprotein Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-44178-0.05ML
This item is not returnable.
View return policy
Description
Gag-Pro-Pol polyprotein Polyclonal specifically detects Gag-Pro-Pol polyprotein in Mouse samples. It is validated for Western Blot, ELISA.
Specifications
| Gag-Pro-Pol polyprotein | |
| Polyclonal | |
| Unconjugated | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% sodium azide | |
| gag-pro-pol | |
| Gag-Pro-Pol polyprotein was prepared from whole rabbit serum produced by repeated immunizations with a full length sequence for mouse mammary tumor virus capsid protein tagged with His. | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 77 mg/mL | |
| Western Blot 1:1000-1:2000, ELISA 1:10000-1:20000 | |
| Capsid protein p27, Gag-Pro-Pol, MMTV capsid, Mouse mammary tumor virus (strain C3H), Reverse transcriptase/ribonuclease H | |
| Rabbit | |
| 0.05 mL | |
| Mouse |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction