missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABA-AR alpha 5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
280.00€ - 624.00€
Specifications
| Antigen | GABA-AR alpha 5 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18249733
|
Novus Biologicals
NBP2-57683 |
100 μL |
624.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18690297
|
Novus Biologicals
NBP2-57683-25ul |
25 μL |
280.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GABA-AR alpha 5 Polyclonal specifically detects GABA-AR alpha 5 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| GABA-AR alpha 5 | |
| Polyclonal | |
| Rabbit | |
| Cellular Markers, Neuronal Cell Markers, Neurotransmission | |
| GABA(A) receptor subunit alpha-5, gamma-aminobutyric acid (GABA) A receptor, alpha 5, gamma-aminobutyric acid receptor subunit alpha-5, MGC138184 | |
| GABRA5 | |
| IgG | |
| Affinity Purified |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| 2558 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:FGFSQMPTSSVKDETNDNITIFT | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title