missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABA-A R beta 1 Antibody (S96-55), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-48319-0.025mg
This item is not returnable.
View return policy
Description
GABA-A R beta 1 Monoclonal antibody specifically detects GABA-A R beta 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Microarray
Specifications
| GABA-A R beta 1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1-10 μg/mL, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 0.1-1.0 μg/mL, Microarray | |
| P50571 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Microarray | |
| S96-55 | |
| Unconjugated | |
| PBS (pH 7.4), 50% Glycerol | |
| GABA(A) receptor subunit beta-1, gamma-aminobutyric acid (GABA) A receptor, beta 1, gamma-aminobutyric acid receptor subunit beta-1 | |
| Fusion protein amino acids 327-450 of mouse GABA-A-R-Beta 1 | |
| 0.025 mg | |
| Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 2560 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction