missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Concentration | 100 mg/mL |
| For Use With (Application) | Resistance to G418 is conferred by the neo gene from transposon Tn5 encoding an aminoglycoside 3'-phosphotransferase, APH 3' II. This protein inactivates G418 by covalently modifying its amino or hydroxyl functions therefore inhibiting the antibiotic-ribosome interaction. |
| Content And Storage | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Quantity | 0.5 g |
| Product Type | G418 |
| Product Line | Novus |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?