Learn More
G3BP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Product Code. 16831084 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16831084 150 μL 150µL
16841084 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16831084 Supplier Proteintech Supplier No. 130572AP150UL
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Rabbit Polyclonal Antibody
May be a regulated effector of stress granule assembly. Phosphorylation-dependent sequence-specific endoribonuclease in vitro. Cleaves exclusively between cytosine and adenine and cleaves c-myc mRNA preferentially at the 3′UTR. ATP- and magnesium-dependen
Specifications
Antigen G3BP1
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene G3BP1
Gene Accession No. P97855, Q13283
Gene Alias G3BP, G3BP 1, G3BP1, HDH VIII
Gene Symbols G3bp1
Host Species Rabbit
Immunogen G3BP1 Fusion Protein Ag3728
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10146, 171092, 27041
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG