Learn More
G3BP1 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1E4A2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66486-1-IG-150UL
This item is not returnable.
View return policy
Description
May be a regulated effector of stress granule assembly. Phosphorylation-dependent sequence-specific endoribonuclease in vitro. Cleaves exclusively between cytosine and adenine and cleaves c-myc mRNA preferentially at the 3′UTR. ATP- and magnesium-dependenSpecifications
| G3BP1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P97855, Q13283 | |
| G3bp1 | |
| G3BP1 Fusion Protein ag3728 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Pig, Rat | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 1E4A2 | |
| Unconjugated | |
| G3BP1 | |
| G3BP, G3BP 1, G3BP1, HDH VIII | |
| Mouse | |
| Protein G | |
| RUO | |
| 100513766, 10146, 171092, 27041 | |
| -20°C | |
| Liquid |