missing translation for 'onlineSavingsMsg'
Learn More
Learn More
G gamma7 Antibody (OTI3G1), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70838AF594
This item is not returnable.
View return policy
Description
G gamma7 Monoclonal specifically detects G gamma7 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| G gamma7 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| GNG7 | |
| Recombinant protein fragment corresponding to amino acids 2-65 of human G gamma7 (NP_443079) produced in E.coli. | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
| Western Blot | |
| OTI3G1 | |
| Western Blot | |
| FLJ00058, GNGT7, guanine nucleotide binding protein (G protein), gamma 7, guanine nucleotide-binding protein G(I)/G(S)/G(O) subunit gamma-7 | |
| Mouse | |
| 0.1 mL | |
| 2788 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction