missing translation for 'onlineSavingsMsg'
Learn More
Learn More
G-CSF Antibody (7E4F7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
217.00€ - 552.00€
Specifications
| Antigen | G-CSF |
|---|---|
| Clone | 7E4F7 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, CyTOF-ready |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18178719
|
Novus Biologicals
NBP2-52447 |
0.1 mg |
552.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18663828
|
Novus Biologicals
NBP2-52447-0.025mg |
0.025 mg |
217.00€
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
G-CSF Monoclonal specifically detects G-CSF in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
| G-CSF | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
| Unconjugated | |
| Mouse | |
| Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research | |
| PBS with 0.05% Sodium Azide | |
| 1440 | |
| Purified recombinant fragment of human CSF3 (AA: 1-207) expressed in E. Coli. | |
| Primary | |
| 22.3 kDa |
| 7E4F7 | |
| Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| C17orf33, chromosome 17 open reading frame 33, colony stimulating factor 3 (granulocyte), CSF3OS, filgrastim, G-CSF, GCSFlenograstim, granulocyte colony-stimulating factor, Lenograstim, MGC45931, pluripoietin | |
| CSF3 | |
| IgG2a | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title