missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FXR1 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67813-1-IG-20UL
This item is not returnable.
View return policy
Description
The FXR1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human FXR1. This antibody recognizes human, mouse, rat antigen. The FXR1 antibody has been validated for the following applications: WB, IHC, IF/ICC, IF-P, IP, ELISA analysis.Specifications
| FXR1 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 8087 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunofluorescence, Immunoprecipitation, ELISA | |
| 1E12E8 | |
| Unconjugated | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 70-80 kDa | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction