Learn More
Equip Labo™ Fume Hood
Product Code. 12516756
Change view
Click to view available options
Dimensions (L x W x H):
180 x 80 x 185 cm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) unitSize
12516756 180 x 80 x 185 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12516756 Supplier Equip Labo™ Supplier No. 2678
  • ONLINE EXCLUSIVE PRICE
    10165.00€
    8508.58€ / Pack of 1
    Save 1656.42€
    16% Off
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
Provides protection against intensive use of acids and other corrosive products in the laboratory.
  • Rear box is equipped with 2 inclined polypropylene panels directing the vapors down
  • A washing device sprays water in the upper part of the box to condense the vapors and avoid the deposition of residues
  • Air dryer at the outlet of the fume hood protects the circuit ventilation
Specifications
Product Type Fume Hood
Dimensions (L x W x H) 180 x 80 x 185 cm