missing translation for 'onlineSavingsMsg'
Learn More

FTO Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

369.00€ - 593.00€

Specifications

Antigen FTO
Dilution Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18641489
View Documents
Novus Biologicals
NBP2-62659-25ul
25 μL
369.00€
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18636667
View Documents
Novus Biologicals
NBP2-62659
100 μg
593.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

FTO Polyclonal antibody specifically detects FTO in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

FTO
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Lipid and Metabolism, Neuroscience
PBS (pH 7.2) and 40% Glycerol
79068
IgG
Protein A purified
Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500
Polyclonal
Purified
RUO
Human
alpha-ketoglutarate-dependent dioxygenase FTO, EC 1.14.11.-, fat mass and obesity associated, Fat mass and obesity-associated protein, KIAA1752protein fto, MGC5149
This antibody was developed against a recombinant protein corresponding to amino acids: LFRDLVRIQGKDLLTPVSRILIGNPGCTYKYLNTRLFTVPWPVKGSNIKHTEAEIAAACETFLKLNDYLQIETIQALEELAA
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.