missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FSTL4 Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA5148926
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
FSTL4 (Follistatin Like 4) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene includecalcium ion binding. An important paralog of this gene isFSTL5.
Specifications
| FSTL4 | |
| Polyclonal | |
| Unconjugated | |
| FSTL4 | |
| B230374F23Rik; follistatin like 4; follistatin-like 4; follistatin-like protein 4; Follistatin-related protein 4; FSL4; FSTL4; KIAA1061; m-D/Bsp120I 1-1; mKIAA1061; SPIG1 | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 23105 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q6MZW2 | |
| FSTL4 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Follistatin-like 4/FSTL4, Trp23-Val842. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction