missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
FSH beta Monoclonal specifically detects FSH beta in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | FSHb/1062 |
| Gene Accession No. | P01225, P01225 |
| Gene Symbols | FSHB |
| Quantity | 0.02 mg |
| Regulatory Status | RUO |
| Content And Storage | Store at 4C. |
| Form | Purified |
| Isotype | IgG1 κ |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?