Learn More
FRMD5 Antibody (CL9503) [DyLight 650], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-88935C
Product Code. 30305421
  • 737.00€ / 0.10mL
Description
FRMD5 Monoclonal antibody specifically detects FRMD5 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FRMD5
Monoclonal
DyLight 650
FERM domain containing 5, FERM domain-containing protein 5, FLJ41022, MGC14161
This antibody was developed using a recombinant protein derived from Q7Z6J6, with the exact immunogen sequence remaining proprietary.
0.1 mL
Primary
Human
Purified
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL9503
50mM Sodium Borate
Mouse
Protein A purified
RUO
84978
Store at 4⪚C in the dark.
IgG2a