missing translation for 'onlineSavingsMsg'
Learn More

Fractin Antibody, Novus Biologicals™

Product Code. 18195955 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18195955 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18195955 Supplier Novus Biologicals Supplier No. NBP229504

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Fractin Polyclonal specifically detects Fractin in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fractin
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:100
Formulation Neat whole antisera with No Preservative
Gene Alias Anti-Fractin fragment of actin
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acid residues from the C terminal region of the 32-kDa actin fragment
Molecular Weight of Antigen 32 kDa
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Cellular Signaling, Neurodegeneration
Primary or Secondary Primary
Test Specificity Specific for the ∼ 32 kDa fractin protein in Western blots with no reactivity to intact actin. There is often a ladder of smaller bands in cells or culture or in vivo preparations due to further degradation by other proteases.
Target Species Human, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.