missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FRA2 Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00002355-B01P
This item is not returnable.
View return policy
Description
FRA2 Polyclonal antibody specifically detects FRA2 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| FRA2 | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/ Immunofluorescence | |
| NP_005244.1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| PBS (pH 7.4) | |
| FOS-like antigen 2, fos-related antigen 2, FRA-2, FRA2FLJ23306 | |
| FOSL2 (NP_005244.1, 1 a.a. - 326 a.a.) full-length human protein. MYQDYPGNFDTSSRGSSGSPAHAESYSSGGGGQQKFRVDMPGSGSAFIPTINAITTSQDLQWMVQPTVITSMSNPYPRSHPYSPLPGLASVPGHMALPRPGVIKTIGTTVGRRRRDEQLSPEEEEKRRIRRERNKLAAAKCRNRRRELTEKLQAETEELEEEKSGLQKEIAELQKEKEKLEFMLVAHGPVCKISPEERRSPPAPGLQPMRSGGGSVGAVVVKQEPLEEDSPSSSSAGLDKAQRSVIKPISIAGGFYGEEPLHTPIVVTSTPAVTPGTSNLVFTYPSVLEQESPASPSESCSKAHRRSSSSGDQSSDSLNSPTLLAL | |
| 0.05 mg | |
| Cancer, Transcription Factors and Regulators, Tumor Suppressors | |
| 2355 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction