missing translation for 'onlineSavingsMsg'
Learn More

FPRL2/FPR3 Antibody (374822), HRP, Novus Biologicals™

Product Code. 18697503 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18697503 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18697503 Supplier Novus Biologicals Supplier No. FAB3896H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

FPRL2/FPR3 Monoclonal antibody specifically detects FPRL2/FPR3 in Human samples. It is validated for Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen FPRL2/FPR3
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 374822
Conjugate HRP
Dilution Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias FMLP-R-II, FMLPY, formyl peptide receptor 3, Formyl peptide receptor-like 2FMLP-related receptor II, FPRH1FPRH2, FPRL2FML2_HUMAN, N-formyl peptide receptor 3, RMLP-R-I
Host Species Mouse
Immunogen HEK293 human embryonic kidney cell line transfected with human FPRL2, Met1-Met352, Accession # P25089
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 2359
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.