Learn More
FoxP4 Antibody (RAB-S40) [Janelia Fluor™ 585], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NBP3-20153JF585
Product Code. 30171743
  • 465.00€ / 0.10mL
Description
FoxP4 Monoclonal antibody specifically detects FoxP4 in Human samples. It is validated for Flow Cytometry,ELISA,Immunoprecipitation,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FoxP4
Monoclonal
Janelia Fluor 585
FKHLA, FLJ40908, FLJ44184, fork head-related protein like A, Fork head-related protein-like A, forkhead box P4, forkhead box protein P4, hFKHLA, winged-helix repressor FOXP4
This antibody was obtained by recombinant antibody (rAb) phage display recognizing FOXP4 protein under non-denaturing conditions.
0.1 mL
Primary
Human
Purified
Flow Cytometry, ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence
RAB-S40
50mM Sodium Borate
Rabbit
Protein A purified
RUO
116113
Store at 4°C in the dark.
IgG κ