missing translation for 'onlineSavingsMsg'
Learn More

FOXO4, p Ser197 Antibody - Azide and BSA Free, Novus Biologicals™

Product Code. 18640647 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18640647 20 μg 20µL
18646027 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18640647 Supplier Novus Biologicals Supplier No. NBP30539320ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FOXO4 Polyclonal antibody specifically detects FOXO4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXO4
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:500, Immunocytochemistry/ Immunofluorescence 1:100 - 1:200
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias AFX, AFX1myeloid/lymphoid or mixed-lineage leukemia (trithorax (Drosophila) homolog);translocated to, 7, Fork head domain transcription factor AFX1, forkhead box O4, forkhead box protein O4, MLLT7MGC120490, myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila);translocated to, 7
Host Species Rabbit
Immunogen A synthetic phosphorylated peptide around S197 of human FOXO4 (NP_005929.2). AASMD
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 4303
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.