missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXO4 Monoclonal Antibody (OTI8F1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810532
This item is not returnable.
View return policy
Description
FOXO4 (Forkhead box O4) is encoded by the FOXO4 gene located on the X chromosome. It is also known as AFX1 or MLLT7 (Myeloid/Lymphoid or Mixed-Lineage Leukemia (Trithorax Homolog, Translocated To, 7) and is a member of the O class of forkhead/ winged helix family of transcription factors along with FOXO1, FOXO3 and FOXO6. FOXO4 is abundantly expressed in skeletal muscle and adipose tissue. FOXO4 binds to the Insulin Responsive Element (IRE) and activates downstream targets of Insulin/IGF1 signaling such as IGFBP1. However, the activation of another downstream target of the Insulin/IGF1 pathway, Akt, leads to phosphorylation of FOXO4 on specific Ser/Thr residues. This phosphorylation mediates binding of FOXO4 to 14-3-3 and eventual exit of FOXO4 from the nucleus.Specifications
| FOXO4 | |
| Monoclonal | |
| Unconjugated | |
| FOXO4 | |
| AFX, AFX1, MLLT7 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 4303 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI8F1 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P98177 | |
| FOXO4 | |
| Human recombit protein fragment corresponding to amino acids 206-505 of human FOXO4 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction