missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXO4 Monoclonal Antibody (OTI10F12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808604S
This item is not returnable.
View return policy
Description
FOXO4 Monoclonal antibody specifically detects FOXO4 in Human samples. It is validated for Western BlotSpecifications
| FOXO4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O75144 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 23308 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI10F12 | |
| Unconjugated | |
| ICOSLG | |
| AFX; Afx1; Afxh; AFXzeta; Fkhr3; Fork head domain transcription factor AFX1; forkhead box O4; forkhead box protein O4; forkhead protein; FOXO4; FOXO4b; MGC120490; MLLT7; myeloid/lymphoid or mixed lineage-leukemia translocation to 7 homolog; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 7; OTTHUMP00000023497; RGD1561201 | |
| Full length human recombinant protein of human ICOSLG produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction